missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorophenylacetic acid, 98%

Catalog No. AAB2135422
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2135422 100 g
AAB2135406 5 g
AAB2135414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2135422 Supplier Thermo Scientific Chemicals Supplier No. B2135422
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6575-24-2 | C8H6Cl2O2 | 205.034 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6575-24-2
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.034
MDL Number MFCD00004320
InChI Key SFAILOOQFZNOAU-UHFFFAOYSA-N
Synonym 2,6-dichlorophenylacetic acid, 2-2,6-dichlorophenyl acetic acid, 2,6-dichlorophenyl acetic acid, benzeneacetic acid, 2,6-dichloro, 2,6-dichloro benzeneacetic acid, 2,6-dichlorophenylaceticacid, unii-k8g025cx8w, 2,6-dichlorobenzeneaceticacid, 2cl
PubChem CID 81058
IUPAC Name 2-(2,6-dichlorophenyl)acetic acid
SMILES C1=CC(=C(C(=C1)Cl)CC(=O)O)Cl

Specifications

Melting Point 158°C to 161°C
Quantity 100 g
Beilstein 1952744
Formula Weight 205.04
Percent Purity 98%
Chemical Name or Material 2,6-Dichlorophenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 229-504-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only