missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloroindophenol, sodium salt hydrate, 90+%

Catalog No. AC450701000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC450701000 100 g Glass bottle
AC450700050 5 g Glass bottle
AC450700250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC450701000 Supplier Thermo Scientific Chemicals Supplier No. 450701000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1266615-56-8 | C12H6Cl2NNaO2 | 290.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1266615-56-8
Molecular Formula C12H6Cl2NNaO2
Molecular Weight (g/mol) 290.07
MDL Number MFCD00150014
InChI Key CVSUAFOWIXUYQA-UHFFFAOYSA-M
Synonym 2,6-dichloroindophenol sodium salt dihydrate, 2,6-dichloro-4-4-hydroxyphenyl azamethylene cyclohexa-2,5-dien-1-one, oxamet hane, oxamethane, sodium salt, c12h6cl2no2.na.2h2o, 2,6-dichlorophenol-indophenol sodium salt dihydrate, sodium 4-3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene amino phenolate dihydrate, sodium 4-3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene amino benzenolate dihydrate
PubChem CID 23696612
IUPAC Name sodium;4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]phenolate;dihydrate
SMILES [Na+].[O-]C1=CC=C(C=C1)N=C1C=C(Cl)C(=O)C(Cl)=C1

Specifications

Assay Percent Range 90+%
Packaging Glass bottle
Quantity 100 g
Solubility Information Solubility in water: soluble. Other solubilities: freely soluble in alcohol
Formula Weight 290.07
Percent Purity 90+%
Chemical Name or Material 2, 6-Dichloroindophenol, sodium salt hydrate

RUO – Research Use Only