missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorodiphenylamine 98.0+%, TCI America™

Catalog No. D377825G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D377825G 25 g
D3778250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D377825G Supplier TCI America Supplier No. D377825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15307-93-4
Molecular Formula C12H9Cl2N
Molecular Weight (g/mol) 238.11
MDL Number MFCD00269648
InChI Key HDUUZPLYVVQTKN-UHFFFAOYSA-N
Synonym 2,6-dichlorodiphenylamine, n-phenyl-2,6-dichloroaniline, benzenamine, 2,6-dichloro-n-phenyl, 2,6-dichlorophenyl phenylamine, dichlorophenylaniline, 2,6-dichlorophenylaniline, diphenylamine,2,6-dichloro, n-phenyl 2,6-dichloroaniline, n-2,6-dichlorophenyl aniline, #
PubChem CID 84867
IUPAC Name 2,6-dichloro-N-phenylaniline
SMILES ClC1=CC=CC(Cl)=C1NC1=CC=CC=C1

Specifications

Melting Point 51°C
Quantity 25 g
Formula Weight 238.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichlorodiphenylamine
EINECSNumber (3)-0061
TSCA No