missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-4-nitroanisole 98,0+%, TCI America™

Numéro de catalogue. D28721G
Modifier l'affichage
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
D28721G 1 g
D28725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. D28721G Fournisseur TCI America Code fournisseur D28721G

Identifiants chimiques

CAS 17742-69-7
Molecular Formula C7H5Cl2NO3
Molecular Weight (g/mol) 222.021
MDL Number MFCD00061129
InChI Key GJYVJKPFYCKNEC-UHFFFAOYSA-N
Synonym 2,6-dichloro-4-nitroanisole, 2,6-dichloro-4-nitro-anisole, 2,6-dichloro-4-nitrophenyl methyl ether, benzene, 1,3-dichloro-2-methoxy-5-nitro, pubchem2687, acmc-1c7kz, gjyvjkpfycknec-uhfffaoysa, 1,3-dichloro-2-methoxy-5-nitrobe, 3,5-dichloro-4-methoxynitrobenzene, 1,3-dichloro-2-methoxy-5-nitro-benzene
PubChem CID 309716
IUPAC Name 1,3-dichloro-2-méthoxy-5-nitrobenzène
SMILES COC1=C(C=C(C=C1Cl)[N+](=O)[O-])Cl

Spécifications

Melting Point 100°C
Quantity 1 g
UN Number 2811
Formula Weight 222.02
Percent Purity ≥98.0% (GC)
Physical Form Poudre cristalline
Chemical Name or Material 2,6-Dichloro-4-nitroanisole
EINECSNumber (3)-4110
TSCA No