missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 110°C |
| Color | White |
| Boiling Point | 103°C |
| Quantity | 5 g |
| Formula Weight | 288.96 |
| Percent Purity | ≥97.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,6-Dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol |
Safety and Handling
| TSCA | No |