missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromonaphthalene-1,4,5,8-tetracarboxylic Dianhydride 98.0+%, TCI America™

Catalog No. D43395G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43395G 5 g
D43391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43395G Supplier TCI America Supplier No. D43395G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 83204-68-6
Molecular Formula C14H2Br2O6
Molecular Weight (g/mol) 425.972
MDL Number MFCD00406909
InChI Key FEDCHNPLDKDTNS-UHFFFAOYSA-N
PubChem CID 11974609
SMILES C1=C2C3=C4C(=CC(=C3C(=O)OC2=O)Br)C(=O)OC(=O)C4=C1Br

Specifications

Melting Point 350°C
Color Yellow
Quantity 5 g
Formula Weight 425.97
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dibromonaphthalene-1,4,5,8-tetracarboxylic Dianhydride