missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromo-N,N'-di-n-octyl-1,8:4,5-naphthalenetetracarboxdiimide 98.0+%, TCI America™

Catalog No. D49465G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49465G 5 g
D49461G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49465G Supplier TCI America Supplier No. D49465G

Chemical Identifiers

CAS 926643-78-9
Molecular Formula C30H36Br2N2O4
Molecular Weight (g/mol) 648.44
MDL Number MFCD29472527
InChI Key HFLBFDNZMQEBCD-UHFFFAOYSA-N
PubChem CID 71623604
IUPAC Name 2,9-dibromo-6,13-dioctyl-6,13-diazatetracyclo[6.6.2.0⁴,¹⁶.0¹¹,¹⁵]hexadeca-1,3,8,10,15-pentaene-5,7,12,14-tetrone
SMILES CCCCCCCCN1C(=O)C2=CC(Br)=C3C(=O)N(CCCCCCCC)C(=O)C4=CC(Br)=C(C1=O)C2=C34

Specifications

Quantity 5 g
Formula Weight 648.44
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dibromo-N,N′-di-n-octyl-1,8:4,5-naphthalenetetracarboxdiimide