missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromo-N,N'-bis(2-ethylhexyl)-1,8:4,5-naphthalenetetracarboxdiimide 98.0+%, TCI America™

Catalog No. D49455G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49455G 5 g
D49451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49455G Supplier TCI America Supplier No. D49455G

Chemical Identifiers

CAS 1088205-02-0
Molecular Formula C30H36Br2N2O4
Molecular Weight (g/mol) 648.44
MDL Number MFCD27923032
InChI Key IMWDVKCSASRBSW-UHFFFAOYNA-N
PubChem CID 71721476
IUPAC Name 2,9-dibromo-6,13-bis(2-ethylhexyl)-6,13-diazatetracyclo[6.6.2.0⁴,¹⁶.0¹¹,¹⁵]hexadeca-1,3,8,10,15-pentaene-5,7,12,14-tetrone
SMILES CCCCC(CC)CN1C(=O)C2=CC(Br)=C3C(=O)N(CC(CC)CCCC)C(=O)C4=CC(Br)=C(C1=O)C2=C34

Specifications

Melting Point 245°C
Quantity 5 g
Formula Weight 648.44
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dibromo-N,N′-bis(2-ethylhexyl)-1,8:4,5-naphthalenetetracarboxdiimide