missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromo-4-n-octyldithieno[3,2-b:2',3'-d]pyrrole 94.0+%, TCI America™

Catalog No. D4714200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4714200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4714200MG Supplier TCI America Supplier No. D4714200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1346688-54-7
Molecular Formula C16H19Br2NS2
Molecular Weight (g/mol) 449.263
InChI Key IJEKMGUFKKPWLT-UHFFFAOYSA-N
PubChem CID 68212207
IUPAC Name 2,6-dibromo-4-octyldithieno[3,2-d
SMILES CCCCCCCCN1C2=C(C3=C1C=C(S3)Br)SC(=C2)Br

Specifications

Color Yellow
Quantity 200 mg
Formula Weight 449.26
Percent Purity ≥94.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dibromo-4-n-octyldithieno[3,2-b:2′,3′-d]pyrrole