missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diaminopyridine, 98%

Catalog No. AAA1229522
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1229522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1229522 Supplier Thermo Scientific Chemicals Supplier No. A1229522
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 141-86-6 | C5H7N3 | 109.132 g/mol

2,6-Diaminopyridine, used as a pharmaceutical intermediate and a hair dye coupler in oxidation/permanent formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Diaminopyridine, used as a pharmaceutical intermediate and a hair dye coupler in oxidation/permanent formulations.

Solubility
Soluble in water 180 g/L @ 20°C, acetone, ethanol, methanol, isopropanol, ethyl acetate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 141-86-6
Molecular Formula C5H7N3
Molecular Weight (g/mol) 109.132
MDL Number MFCD00006329
InChI Key VHNQIURBCCNWDN-UHFFFAOYSA-N
Synonym 2,6-diaminopyridine, 2,6-pyridinediamine, 2,6-diamino pyridine, pyridine, 2,6-diamino, dap amine, pyridine-2,6-diyldiamine, unii-k89ab8ckg6, ccris 6682, k89ab8ckg6, dsstox_cid_20127
PubChem CID 8861
IUPAC Name pyridine-2,6-diamine
SMILES C1=CC(=NC(=C1)N)N

Specifications

Melting Point 118°C to 121°C
Boiling Point 288°C
Flash Point 155°C (311°F)
Odor Unpleasant
Quantity 100 g
UN Number UN2671
Beilstein 108513
Solubility Information Soluble in water 180g/L @ 20°C,acetone,ethanol,methanol,isopropanol,ethyl acetate.
Formula Weight 109.13
Percent Purity 98%
Chemical Name or Material 2,6-Diaminopyridine
Show More Show Less
Hazard Category H301-H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H373-H319-H335
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: AMINOPYRIDINES
EINECSNumber 205-507-2
RTECSNumber US7570000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only