missing translation for 'onlineSavingsMsg'
Learn More

2,6-Di-tert-butyl-4-methylphenol, 99%

Catalog No. AAA168630B
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA168630B 1000 g
AAA1686330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA168630B Supplier Thermo Scientific Chemicals Supplier No. A168630B
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 128-37-0 | C15H24O | 220.356 g/mol

2,6-Di-tert-butyl-4-methylphenol is used as an antioxidant for cosmetics, vitamins, pharmaceuticals, rubber, oils and fats. It is used as a fuel additive in the petroleum industry and also used in hydraulic fluids, turbine and gear oils and jet fuels. It acts as a stabilizer in diethyl ether, tetrahydrofuran and other laboratory chemicals to prevent peroxide formation. It is effectively involved as a polymerization inhibitor in the process of oxidation of allyl alcohol to glycerine. It is utilized in the synthesis of organoaluminum compound, methylaluminum bis(2,6-di-tert-butyl-4-alkylphenoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Di-tert-butyl-4-methylphenol is used as an antioxidant for cosmetics, vitamins, pharmaceuticals, rubber, oils and fats. It is used as a fuel additive in the petroleum industry and also used in hydraulic fluids, turbine and gear oils and jet fuels. It acts as a stabilizer in diethyl ether, tetrahydrofuran and other laboratory chemicals to prevent peroxide formation. It is effectively involved as a polymerization inhibitor in the process of oxidation of allyl alcohol to glycerine. It is utilized in the synthesis of organoaluminum compound, methylaluminum bis(2,6-di-tert-butyl-4-alkylphenoxide.

Solubility
Soluble in methanol, toluene, isopropanol, methyl ethyl ketone, acetone, benzene, petroleum ether and ethanol. Insoluble in water.

Notes
Light-sensitive. Keep the container tightly closed in a cool, dry and well-ventilated place. Incompatible with acid chlorides, acid anhydrides, oxidizing agents, bases, brass, copper and copper alloys.

Chemical Identifiers

CAS 128-37-0
Molecular Formula C15H24O
Molecular Weight (g/mol) 220.356
MDL Number MFCD00011644
InChI Key NLZUEZXRPGMBCV-UHFFFAOYSA-N
Synonym 2,6-di-tert-butyl-4-methylphenol, butylated hydroxytoluene, butylhydroxytoluene, 2,6-di-tert-butyl-p-cresol, 2,6-di-t-butyl-4-methylphenol, ionol, dbpc, dibunol, stavox, bht
PubChem CID 31404
ChEBI CHEBI:34247
IUPAC Name 2,6-ditert-butyl-4-methylphenol
SMILES CC1=CC(=C(C(=C1)C(C)(C)C)O)C(C)(C)C

Specifications

Melting Point 69°C to 72°C
Density 1.048
Boiling Point 264°C to 265°C
Flash Point 127°C (260°F)
Odor Mild
Quantity 1000 g
UN Number UN3077
Beilstein 1911640
Merck Index 14,1548
Solubility Information Soluble in methanol,toluene,isopropanol,methyl ethyl ketone,acetone,benzene,petroleum ether and ethanol. Insoluble in water.
Formula Weight 220.36
Percent Purity 99%
Chemical Name or Material 2,6-Di-tert-butyl-4-methylphenol
Show More Show Less
Hazard Category H335-H373
Hazard Statement GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement P260-P271-P304+P340-P312-P314-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 204-881-4
RTECSNumber GO7875000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only