missing translation for 'onlineSavingsMsg'
Learn More

2,6-Di-tert-butyl-4-hydroxymethylphenol 97.0+%, TCI America™

Catalog No. D148025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D148025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D148025G Supplier TCI America Supplier No. D148025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 88-26-6
Molecular Formula C15H24O2
Molecular Weight (g/mol) 236.355
MDL Number MFCD00017254
InChI Key HNURKXXMYARGAY-UHFFFAOYSA-N
Synonym 3,5-di-tert-butyl-4-hydroxybenzyl alcohol, 2,6-di-tert-butyl-4-hydroxymethyl phenol, 2,6-di-tert-butyl-4-hydroxymethylphenol, bht alcohol, antioxidant 754, ionox 100, ionox 100 antioxidant, 3,5-di-t-butyl-4-hydroxybenzyl alcohol, butylated hydroxymethylphenol, 2,6-di-t-butyl-4-hydroxymethylphenol
PubChem CID 6929
IUPAC Name 2,6-ditert-butyl-4-(hydroxymethyl)phenol
SMILES CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)CO

Specifications

Melting Point 140°C
Quantity 25 g
Formula Weight 236.36
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Di-tert-butyl-4-hydroxymethylphenol
EINECSNumber (3)-0542&(3)-1079
RTECSNumber DO0750000
TSCA Yes