missing translation for 'onlineSavingsMsg'
Learn More

2,6-Bis(trifluoromethyl)benzeneboronic acid, 97%

CAS: 681812-07-7 | C8H5BF6O2 | 257.926 g/mol

$132.04 - $534.89

Chemical Identifiers

CAS 681812-07-7
Molecular Formula C8H5BF6O2
Molecular Weight (g/mol) 257.926
MDL Number MFCD04039321
InChI Key WAMPGNNEOZGBHR-UHFFFAOYSA-N
Synonym 2,6-bis trifluoromethyl phenyl boronic acid, 2,6-bis trifluoromethyl phenylboronic acid, 2,6-bis trifluoromethyl benzeneboronic acid, 2,6-ditrifluoromethylphenyl boronic acid, boronic acid, 2,6-bis trifluoromethyl phenyl, acmc-1b92w, ablock ab-13-3574, 2,6-bis-trifluoromethyl benzeneboronic acid
PubChem CID 2783239
IUPAC Name [2,6-bis(trifluoromethyl)phenyl]boronic acid
SMILES B(C1=C(C=CC=C1C(F)(F)F)C(F)(F)F)(O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL19952MD
View Documents
Thermo Scientific Chemicals
L19952MD
250 mg
Each for $132.04
Only null left
 
AAL1995203
View Documents
Thermo Scientific Chemicals
L1995203
1 g
Each for $160.12
Only null left
 
AAL1995206
View Documents
Thermo Scientific Chemicals
L1995206
5 g
Each for $534.89
Only null left
 
Description

Description

2,6-Bis(trifluoromethyl)benzeneboronic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Bis(trifluoromethyl)benzeneboronic acid is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

681812-07-7
257.926
WAMPGNNEOZGBHR-UHFFFAOYSA-N
2783239
B(C1=C(C=CC=C1C(F)(F)F)C(F)(F)F)(O)O
C8H5BF6O2
MFCD04039321
2,6-bis trifluoromethyl phenyl boronic acid, 2,6-bis trifluoromethyl phenylboronic acid, 2,6-bis trifluoromethyl benzeneboronic acid, 2,6-ditrifluoromethylphenyl boronic acid, boronic acid, 2,6-bis trifluoromethyl phenyl, acmc-1b92w, ablock ab-13-3574, 2,6-bis-trifluoromethyl benzeneboronic acid
[2,6-bis(trifluoromethyl)phenyl]boronic acid

Specifications

681812-07-7
C8H5BF6O2
250 mg
Insoluble in water.
B(C1=C(C=CC=C1C(F)(F)F)C(F)(F)F)(O)O
257.926
257.93
2,6-Bis(trifluoromethyl)benzeneboronic acid
∼150°C
MFCD04039321
2,6-bis trifluoromethyl phenyl boronic acid, 2,6-bis trifluoromethyl phenylboronic acid, 2,6-bis trifluoromethyl benzeneboronic acid, 2,6-ditrifluoromethylphenyl boronic acid, boronic acid, 2,6-bis trifluoromethyl phenyl, acmc-1b92w, ablock ab-13-3574, 2,6-bis-trifluoromethyl benzeneboronic acid
WAMPGNNEOZGBHR-UHFFFAOYSA-N
[2,6-bis(trifluoromethyl)phenyl]boronic acid
2783239
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only