CAS RN 681812-07-7
IUPAC Name:
[2,6-bis(trifluoromethyl)phenyl]boronic acid
Synonyms:
2,6-bis(trifluoromethyl)phenylboronic acid
Molecular Weight (g/mol):
257.93
Molecular Formula:
C8H5BF6O2
InChi Key:
WAMPGNNEOZGBHR-UHFFFAOYSA-N
SMILES:
OB(O)C1=C(C=CC=C1C(F)(F)F)C(F)(F)F
1
–
1
of
1
results
1
Promotions Available
2,6-Bis(trifluoromethyl)benzeneboronic acid, 97%
2,6-Bis(trifluoromethyl)benzeneboronic acid, 97%
CAS: 681812-07-7 Molecular Formula: C8H5BF6O2 Molecular Weight (g/mol): 257.926 MDL Number: MFCD04039321 InChI Key: WAMPGNNEOZGBHR-UHFFFAOYSA-N Synonym: 2,6-bis trifluoromethyl phenyl boronic acid,2,6-bis trifluoromethyl phenylboronic acid,2,6-bis trifluoromethyl benzeneboronic acid,2,6-ditrifluoromethylphenyl boronic acid,boronic acid, 2,6-bis trifluoromethyl phenyl,acmc-1b92w,ablock ab-13-3574,2,6-bis-trifluoromethyl benzeneboronic acid PubChem CID: 2783239 IUPAC Name: [2,6-bis(trifluoromethyl)phenyl]boronic acid SMILES: B(C1=C(C=CC=C1C(F)(F)F)C(F)(F)F)(O)O