Learn More
2,6-Bis(trifluoromethyl)benzeneboronic acid, 97%
CAS: 681812-07-7 | C8H5BF6O2 | 257.926 g/mol
Supplier: Thermo Scientific Chemicals L1995206
Description
2,6-Bis(trifluoromethyl)benzeneboronic acid is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,6-Bis(trifluoromethyl)benzeneboronic acid is used as pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
| 2,6-Bis(trifluoromethyl)benzeneboronic acid | |
| ∼150°C | |
| MFCD04039321 | |
| 2,6-bis trifluoromethyl phenyl boronic acid, 2,6-bis trifluoromethyl phenylboronic acid, 2,6-bis trifluoromethyl benzeneboronic acid, 2,6-ditrifluoromethylphenyl boronic acid, boronic acid, 2,6-bis trifluoromethyl phenyl, acmc-1b92w, ablock ab-13-3574, 2,6-bis-trifluoromethyl benzeneboronic acid | |
| WAMPGNNEOZGBHR-UHFFFAOYSA-N | |
| [2,6-bis(trifluoromethyl)phenyl]boronic acid | |
| 2783239 | |
| 97% |
| 681812-07-7 | |
| C8H5BF6O2 | |
| 5 g | |
| Insoluble in water. | |
| B(C1=C(C=CC=C1C(F)(F)F)C(F)(F)F)(O)O | |
| 257.926 | |
| 257.93 |
Chemical Identifiers
| 681812-07-7 | |
| 257.926 | |
| WAMPGNNEOZGBHR-UHFFFAOYSA-N | |
| 2783239 | |
| B(C1=C(C=CC=C1C(F)(F)F)C(F)(F)F)(O)O |
| C8H5BF6O2 | |
| MFCD04039321 | |
| 2,6-bis trifluoromethyl phenyl boronic acid, 2,6-bis trifluoromethyl phenylboronic acid, 2,6-bis trifluoromethyl benzeneboronic acid, 2,6-ditrifluoromethylphenyl boronic acid, boronic acid, 2,6-bis trifluoromethyl phenyl, acmc-1b92w, ablock ab-13-3574, 2,6-bis-trifluoromethyl benzeneboronic acid | |
| [2,6-bis(trifluoromethyl)phenyl]boronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only