missing translation for 'onlineSavingsMsg'
Learn More

2,6-Bis(2-benzimidazolyl)pyridine 98.0+%, TCI America™

Numéro de catalogue. B35101G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
B35101G 1 g
B35105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. B35101G Fournisseur TCI America Code fournisseur B35101G
Il en reste null

Identifiants chimiques

CAS 28020-73-7
Molecular Formula C19H13N5
Molecular Weight (g/mol) 311.35
MDL Number MFCD00134500
InChI Key JBKICBDXAZNSKA-UHFFFAOYSA-N
PubChem CID 738986
IUPAC Name 2-[6-(1H-1,3-benzodiazol-2-yl)pyridin-2-yl]-1H-1,3-benzodiazole
SMILES N1C2=CC=CC=C2N=C1C1=CC=CC(=N1)C1=NC2=CC=CC=C2N1

Spécifications

Melting Point 353°C
Color White-Yellow
Quantity 1 g
Synonym 2,2′C-(2,6-Pyridinediyl)bis(benzimidazole)
Formula Weight 311.35
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Bis(2-benzimidazolyl)pyridine