missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6,10-Trimethyl-2,6,10-triazaundecane 96.0+%, TCI America™
$65.24 - $413.83
Chemical Identifiers
| CAS | 3855-32-1 |
|---|---|
| Molecular Formula | C11H27N3 |
| Molecular Weight (g/mol) | 201.358 |
| MDL Number | MFCD00126936 |
| InChI Key | SKCNNQDRNPQEFU-UHFFFAOYSA-N |
| Synonym | N-Methyl-N,N-bis[3-(dimethylamino)propyl]amine |
| PubChem CID | 77463 |
| IUPAC Name | N'-[3-(dimethylamino)propyl]-N,N,N'-trimethylpropane-1,3-diamine |
| SMILES | CN(C)CCCN(C)CCCN(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T161825ML
|
TCI America
T161825ML |
25 mL |
Each for $65.24
|
|
|||||
|
T1618500ML
|
TCI America
T1618500ML |
500 mL |
Each for $413.83
|
|
|||||
Chemical Identifiers
| 3855-32-1 | |
| 201.358 | |
| SKCNNQDRNPQEFU-UHFFFAOYSA-N | |
| 77463 | |
| CN(C)CCCN(C)CCCN(C)C |
| C11H27N3 | |
| MFCD00126936 | |
| N-Methyl-N,N-bis[3-(dimethylamino)propyl]amine | |
| N'-[3-(dimethylamino)propyl]-N,N,N'-trimethylpropane-1,3-diamine |
Specifications
| 3855-32-1 | |
| 102°C | |
| MFCD00126936 | |
| 2735 | |
| SKCNNQDRNPQEFU-UHFFFAOYSA-N | |
| N'-[3-(dimethylamino)propyl]-N,N,N'-trimethylpropane-1,3-diamine | |
| 77463 | |
| ≥96.0% (GC,T) | |
| 2,6,10-Trimethyl-2,6,10-triazaundecane |
| Yellow | |
| C11H27N3 | |
| 25 mL | |
| N-Methyl-N,N-bis[3-(dimethylamino)propyl]amine | |
| CN(C)CCCN(C)CCCN(C)C | |
| 201.358 | |
| 201.36 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-3284
TSCA : Yes