accessibility menu, dialog, popup

UserName

CAS RN 3855-32-1

H3CNCH3NCH3NCH3CH3

CAS RN 3855-32-1

IUPAC Name: (3-{[3-(dimethylamino)propyl](methyl)amino}propyl)dimethylamine
Synonyms: (3-{[3-(dimethylamino)propyl](methyl)amino}propyl)dimethylamine
Molecular Weight (g/mol): 201.36
Molecular Formula: C11H27N3
InChi Key: SKCNNQDRNPQEFU-UHFFFAOYSA-N
SMILES: CN(C)CCCN(C)CCCN(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results