missing translation for 'onlineSavingsMsg'
Learn More

2,6,10-Trimethyl-2,6,10-triazaundecane 96.0+%, TCI America™

Supplier:  TCI America T1618500ML

 View more versions of this product

Catalog No. T1618500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2,6,10-Trimethyl-2,6,10-triazaundecane
Yellow
C11H27N3
500 mL
N-Methyl-N,N-bis[3-(dimethylamino)propyl]amine
CN(C)CCCN(C)CCCN(C)C
201.358
201.36
Liquid
3855-32-1
102°C
MFCD00126936
2735
SKCNNQDRNPQEFU-UHFFFAOYSA-N
N'-[3-(dimethylamino)propyl]-N,N,N'-trimethylpropane-1,3-diamine
77463
≥96.0% (GC,T)

Chemical Identifiers

3855-32-1
201.358
SKCNNQDRNPQEFU-UHFFFAOYSA-N
77463
CN(C)CCCN(C)CCCN(C)C
C11H27N3
MFCD00126936
N-Methyl-N,N-bis[3-(dimethylamino)propyl]amine
N'-[3-(dimethylamino)propyl]-N,N,N'-trimethylpropane-1,3-diamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3284

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions