missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6,10-Trimethyl-2,6,10-triazaundecane 96.0+%, TCI America™
Supplier: TCI America T161825ML
Specifications
| 2,6,10-Trimethyl-2,6,10-triazaundecane | |
| Yellow | |
| C11H27N3 | |
| 25 mL | |
| N-Methyl-N,N-bis[3-(dimethylamino)propyl]amine | |
| CN(C)CCCN(C)CCCN(C)C | |
| 201.358 | |
| 201.36 | |
| Liquid |
| 3855-32-1 | |
| 102°C | |
| MFCD00126936 | |
| 2735 | |
| SKCNNQDRNPQEFU-UHFFFAOYSA-N | |
| N'-[3-(dimethylamino)propyl]-N,N,N'-trimethylpropane-1,3-diamine | |
| 77463 | |
| ≥96.0% (GC,T) |
Chemical Identifiers
| 3855-32-1 | |
| 201.358 | |
| SKCNNQDRNPQEFU-UHFFFAOYSA-N | |
| 77463 | |
| CN(C)CCCN(C)CCCN(C)C |
| C11H27N3 | |
| MFCD00126936 | |
| N-Methyl-N,N-bis[3-(dimethylamino)propyl]amine | |
| N'-[3-(dimethylamino)propyl]-N,N,N'-trimethylpropane-1,3-diamine |
Safety and Handling
EINECSNumber : (2)-3284
TSCA : Yes