missing translation for 'onlineSavingsMsg'
Learn More

2,5-Norbornadiene (stabilized with BHT) 97.0+%, TCI America™

Catalog No. N0346500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0346500ML 500 mL
N034625ML 25 mL
N0346100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N0346500ML Supplier TCI America Supplier No. N0346500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 121-46-0
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00082301
InChI Key SJYNFBVQFBRSIB-UHFFFAOYSA-N
Synonym 2,5-norbornadiene, bicyclo 2.2.1 hepta-2,5-diene, norbornadiene, unii-w9ztq75zus, bicyclo 2.2.1 heptadiene, 8,9,10-trinorborna-2,5-diene, w9ztq75zus, dicycloheptadiene
PubChem CID 8473
IUPAC Name bicyclo[2.2.1]hepta-2,5-diene
SMILES C1C2C=CC1C=C2

Specifications

Color Yellow
Boiling Point 90°C
Quantity 500 mL
UN Number 2251
Formula Weight 92.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Norbornadiene (stabilized with BHT)
EINECSNumber (4)-1220
RTECSNumber RB6535000
TSCA Yes