missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylphenylacetonitrile 98.0+%, TCI America™

Catalog No. p-7128189
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42715G 5 g
D427125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42715G Supplier TCI America Supplier No. D42715G
Only null left

Chemical Identifiers

CAS 16213-85-7
Molecular Formula C10H11N
Molecular Weight (g/mol) 145.205
MDL Number MFCD00013816
InChI Key HCELORQTHSKCQL-UHFFFAOYSA-N
Synonym 2,5-dimethylphenylacetonitrile, 2-2,5-dimethylphenyl acetonitrile, 2,5-dimethylbenzyl cyanide, acetonitrile, 2,5-xylyl, 2,5-xylyl acetonitrile, benzeneacetonitrile, 2,5-dimethyl, 2,5-xylylacetonitrile, 2,5-dimethylphenyl acetonitrile, 2,5-dimethylbenzeneacetonitrile, unii-ult00p22mn
PubChem CID 27753
IUPAC Name 2-(2,5-dimethylphenyl)acetonitrile
SMILES CC1=CC(=C(C=C1)C)CC#N

Specifications

Melting Point 30°C
Color White
Boiling Point 79°C
Quantity 5 g
UN Number 3439
Formula Weight 145.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,5-Dimethylphenylacetonitrile
RTECSNumber AM2675000
TSCA No