missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylfuran-3-carbonyl chloride, 97%, Thermo Scientific™

Numéro de catalogue. CC01102DE
Modifier l'affichage
Click to view available options
Quantité:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC01102DE 5g
CC01102DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CC01102DE Fournisseur Thermo Scientific Chemicals Code fournisseur CC01102DE

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 50990-93-7
Formule moléculaire C7H7ClO2
Poids moléculaire (g/mol) 158.58
Numéro MDL MFCD00221073
Clé InChI RWORXZHVOUPMMM-UHFFFAOYSA-N
Synonyme 2,5-dimethyl-3-furoyl chloride, 2,5-dimethyl-furan-3-carbonyl chloride, 2,5-dimethyl-3-furancarbonyl chloride, 2,5-dimethylfuran-3-carbonylchloride, pubchem6936, acmc-209krc, 2,5-dimethyl-3-furoylchloride, 2,5 dimethyl-furan-3-carbonyl chloride, 2,5-dimethyl furan-3-carbonyl chloride, 3-furancarbonylchloride, 2,5-dimethyl
PubChem CID 2734804
Nom de l’IUPAC 2,5-dimethylfuran-3-carbonyl chloride
SOURIRES CC1=CC(C(Cl)=O)=C(C)O1

Spécifications

Nom ou substance chimique 2, 5-Dimethylfuran-3-carbonyl chloride
CAS % minimum 96.5
CAS % maximal 100.0
Densité 1.19g/mL
Fourchette de pourcentages d’essai 97%
Quantité 5g
Poids de formule 158.58
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.