missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-2-hexanol 98.0+%, TCI America™

Catalog No. D19685ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D19685ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D19685ML Supplier TCI America Supplier No. D19685ML

Chemical Identifiers

CAS 3730-60-7
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.23
MDL Number MFCD00021814
InChI Key JPUIYNHIEXIFMV-UHFFFAOYSA-N
Synonym 2,5-dimethyl-2-hexanol, 2-hexanol, 2,5-dimethyl-, s, 2-hexanol, 2,5-dimethyl, 2-hexanol, 2,5-dimethyl-, s-+, acmc-209is7, 2-hexanol,2,5-dimethyl, 2-hexanol,5-dimethyl-, s, 2-hexanol,5-dimethyl-, s-+
PubChem CID 19506
IUPAC Name 2,5-dimethylhexan-2-ol
SMILES CC(C)CCC(C)(C)O

Specifications

Color Colorless
Boiling Point 74°C
Quantity 5 mL
UN Number 1987
Formula Weight 130.23
Physical Form Liquid
Chemical Name or Material 2,5-Dimethyl-2-hexanol
EINECSNumber (2)-0217
TSCA No