missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. D35221G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D35221G Supplier TCI America Supplier No. D35221G

Chemical Identifiers

CAS 107099-99-0
Molecular Formula C8H11BO4
Molecular Weight (g/mol) 181.982
MDL Number MFCD01318181
InChI Key QOZLFNQLIKOGDR-UHFFFAOYSA-N
Synonym 2,5-dimethoxyphenyl boronic acid, 2,5-dimethoxybenzeneboronic acid, 2,5-dimethoxyphenylboronicacid, 2,5-dimethoxyphenyl boranediol, boronic acid, 2,5-dimethoxyphenyl, pubchem1824, acmc-2098nk, ksc174i7n, 2,5-dimethoxyphenylboronoic acid
PubChem CID 2734342
IUPAC Name (2,5-dimethoxyphenyl)boronic acid
SMILES B(C1=C(C=CC(=C1)OC)OC)(O)O

Specifications

Melting Point 97°C
Color White-Yellow
Quantity 1 g
Formula Weight 181.98
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dimethoxyphenylboronic Acid (contains varying amounts of Anhydride)