missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethoxy-1,4-benzoquinone 98.0+%, TCI America™

Catalog No. D22505G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22505G 5 g
D225025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D22505G Supplier TCI America Supplier No. D22505G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3117-03-1
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00189386
InChI Key RMMPZDDLWLALLJ-UHFFFAOYSA-N
Synonym 2,5-Dimethoxy-p-quinone
PubChem CID 101405
IUPAC Name 2,5-dimethoxycyclohexa-2,5-diene-1,4-dione
SMILES COC1=CC(=O)C(=CC1=O)OC

Specifications

Quantity 5 g
Formula Weight 168.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dimethoxy-1,4-benzoquinone