missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 2,5-Dihydro-2,5-dimethoxyfuran, cis + trans, 99%

Catalog No. AAB2502818
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2502818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2502818 Supplier Thermo Scientific™ Supplier No. B2502818

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 332-77-4 | C6H10O3 | 130.14 g/mol

Chemical Identifiers

CAS 332-77-4
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
MDL Number MFCD00003220
InChI Key WXFWXFIWDGJRSC-UHFFFAOYNA-N
Synonym 2,5-Dimethoxy-2,5-dihydrofuran;
IUPAC Name 2,5-dimethoxy-2,5-dihydrofuran
SMILES COC1OC(OC)C=C1

Specifications

Melting Point <-40°
Density 1.072
Boiling Point 160-162°
Flash Point 47°(116°F)
Quantity 50 g
UN Number UN1993
Formula Weight 130.14
Percent Purity 99%
Physical Form Liquid
ShelfLife 3 Years
EINECSNumber 206-367-5