missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluorobenzotrifluoride, 98%

Catalog No. AAH6411106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6411106 5 g
AAH6411103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6411106 Supplier Thermo Scientific Chemicals Supplier No. H6411106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 393-38-4 | C7H3F5 | 182.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 393-38-4
Molecular Formula C7H3F5
Molecular Weight (g/mol) 182.09
MDL Number MFCD00042494
InChI Key BLDOSVXRDMTZQE-UHFFFAOYSA-N
Synonym 2,5-difluorobenzotrifluoride, 1,4-difluoro-2-trifluoromethyl benzene, 1,4-difluoro-2-trifluoromethyl-benzene, benzene, 1,4-difluoro-2-trifluoromethyl, pubchem1650, 1-trifluoromethyl-2,5-difluorobenzene
PubChem CID 11111097
IUPAC Name 1,4-difluoro-2-(trifluoromethyl)benzene
SMILES FC1=CC(=C(F)C=C1)C(F)(F)F

Specifications

Melting Point -14°C
Color Colorless to Yellow
Density 1.424 g/mL
Boiling Point 107°C to 108°C
Refractive Index 1.394
Quantity 5 g
Beilstein 2049494
Formula Weight 182.09
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2,5-Difluorobenzotrifluoride
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only