missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluorobenzonitrile 98.0+%, TCI America™

Catalog No. D18345G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D18345G 5 g
D183425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D18345G Supplier TCI America Supplier No. D18345G

Chemical Identifiers

CAS 64248-64-2
Molecular Formula C7H3F2N
Molecular Weight (g/mol) 139.11
MDL Number MFCD00001777
InChI Key OJTMHIMQUQOLJV-UHFFFAOYSA-N
Synonym benzonitrile, 2,5-difluoro, 2,5-difluoro benzonitrile, 2,5-difluoro-benzonitrile, 2,5-difluorobenzenecarbonitrile, ncr bf ef, pubchem4754, 2,5-diflurobenzonitrile, 2,5 difluorobenzonitrile, 2,5-difluorobenzonitrile
PubChem CID 123558
IUPAC Name 2,5-difluorobenzonitrile
SMILES FC1=CC=C(F)C(=C1)C#N

Specifications

Melting Point 35°C
Color White-Yellow
Boiling Point 92°C
Quantity 5 g
UN Number 3439
Formula Weight 139.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,5-Difluorobenzonitrile