missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorothiophene-3-carbonyl chloride, 95%, Thermo Scientific™

Catalog No. p-2696745
Change view
Click to view available options
Quantity:
1g
10g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CD09458DA 1g Amber Glass Bottle
CD09458EA 10g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CD09458DA Supplier Thermo Scientific Chemicals Supplier No. CD09458DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 57248-14-3
Molecular Formula C5HCl3OS
Molecular Weight (g/mol) 215.472
MDL Number MFCD00041441
InChI Key YGDFSMANOSKQRU-UHFFFAOYSA-N
Synonym 2,5-dichlorothiophene-3-carbonylchloride, 2,5-dichloro-3-thiophenecarbonyl chloride, 3-thiophenecarbonylchloride, 2,5-dichloro, pubchem15832, 2,5-dichloro-3-thenoyl chloride, 2,5-dichloro-3-thiophenecarbonylchloride, 2,5-dichloro-thiophene-3-carbonyl chloride, 2,5-bis chloranyl thiophene-3-carbonyl chloride
PubChem CID 2777084
IUPAC Name 2,5-dichlorothiophene-3-carbonyl chloride
SMILES C1=C(SC(=C1C(=O)Cl)Cl)Cl

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 215.49
Percent Purity 95%
Chemical Name or Material 2, 5-Dichlorothiophene-3-carbonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.