missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorosulfanilic Acid 98.0+%, TCI America™

Catalog No. D0415500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0415500G 500 g
D041525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0415500G Supplier TCI America Supplier No. D0415500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 88-50-6
Molecular Formula C6H5Cl2NO3S
Molecular Weight (g/mol) 242.07
MDL Number MFCD00025159
InChI Key SJCTXIKOXTUQHC-UHFFFAOYSA-N
Synonym 2,5-Dichloroaniline-4-sulfonic Acid
PubChem CID 66619
IUPAC Name 4-amino-2,5-dichlorobenzenesulfonic acid
SMILES C1=C(C(=CC(=C1Cl)S(=O)(=O)O)Cl)N

Specifications

Color White-Yellow
Quantity 500 g
Formula Weight 242.07
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dichlorosulfanilic Acid
EINECSNumber (3)-2085
TSCA Yes