missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloropyrimidine, 95%

Catalog No. AAH6406622
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6406622 100 g
AAH6406606 5 g
AAH6406614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6406622 Supplier Thermo Scientific Chemicals Supplier No. H6406622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 22536-67-0 | C4H2Cl2N2 | 148.974 g/mol

2,5-Dichloropyrimidine acid is employed as a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dichloropyrimidine acid is employed as a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 22536-67-0
Molecular Formula C4H2Cl2N2
Molecular Weight (g/mol) 148.974
MDL Number MFCD00673131
InChI Key CEJAHXLRNZJPQH-UHFFFAOYSA-N
Synonym 2,5-dichloro-pyrimidine, pyrimidine, 2,5-dichloro, 2,5-dichloro-1,3-diazine, pubchem13600, acmc-209fxc, ksc493a8n, 2,5-dichloropyrimidine, pyrimidine, 2,5-dichloro-7ci,8ci,9ci
PubChem CID 590641
IUPAC Name 2,5-dichloropyrimidine
SMILES C1=C(C=NC(=N1)Cl)Cl

Specifications

Melting Point 48°C to 55°C
Color White
Flash Point 107°C (225°F)
Quantity 100 g
UN Number UN3077
Beilstein 110912
Solubility Information Slightly soluble in water.
Formula Weight 148.97
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 2,5-Dichloropyrimidine
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only