missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorobenzotrifluoride 98.0+%, TCI America™

Supplier:  TCI America D13185G

 View more versions of this product

Catalog No. D13185G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2,5-Dichlorobenzotrifluoride
Yellow
MFCD00000608
1760
DYBYUWVMLBBEMA-UHFFFAOYSA-N
1,4-dichloro-2-(trifluoromethyl)benzene
35360
≥98.0% (GC)
320-50-3
C7H3Cl2F3
5 g
2,5-dichlorobenzotrifluoride, 1,4-dichloro-2-trifluoromethyl benzene, benzene, 1,4-dichloro-2-trifluoromethyl, 2,5-dichloro-alpha,alpha,alpha-trifluorotoluene, 2,5-dichloro-a,a,a-trifluorotoluene, toluene, 2,5-dichloro-alpha,alpha,alpha-trifluoro-6ci,7ci,8ci, pubchem3450, ksc925c7r, 2,5-dichloro trifluoromethylbenzene
C1=CC(=C(C=C1Cl)C(F)(F)F)Cl
214.996
215.00
Liquid

Chemical Identifiers

320-50-3
214.996
DYBYUWVMLBBEMA-UHFFFAOYSA-N
35360
C1=CC(=C(C=C1Cl)C(F)(F)F)Cl
C7H3Cl2F3
MFCD00000608
2,5-dichlorobenzotrifluoride, 1,4-dichloro-2-trifluoromethyl benzene, benzene, 1,4-dichloro-2-trifluoromethyl, 2,5-dichloro-alpha,alpha,alpha-trifluorotoluene, 2,5-dichloro-a,a,a-trifluorotoluene, toluene, 2,5-dichloro-alpha,alpha,alpha-trifluoro-6ci,7ci,8ci, pubchem3450, ksc925c7r, 2,5-dichloro trifluoromethylbenzene
1,4-dichloro-2-(trifluoromethyl)benzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : CZ5553755

TSCA : No

SDS
Documents

Documents

Promotions

Promotions