missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorobenzotrifluoride 98.0+%, TCI America™

Catalog No. p-7126673
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D13185G 5 g
D131825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D13185G Supplier TCI America Supplier No. D13185G
Only null left

Chemical Identifiers

CAS 320-50-3
Molecular Formula C7H3Cl2F3
Molecular Weight (g/mol) 214.996
MDL Number MFCD00000608
InChI Key DYBYUWVMLBBEMA-UHFFFAOYSA-N
Synonym 2,5-dichlorobenzotrifluoride, 1,4-dichloro-2-trifluoromethyl benzene, benzene, 1,4-dichloro-2-trifluoromethyl, 2,5-dichloro-alpha,alpha,alpha-trifluorotoluene, 2,5-dichloro-a,a,a-trifluorotoluene, toluene, 2,5-dichloro-alpha,alpha,alpha-trifluoro-6ci,7ci,8ci, pubchem3450, ksc925c7r, 2,5-dichloro trifluoromethylbenzene
PubChem CID 35360
IUPAC Name 1,4-dichloro-2-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1Cl)C(F)(F)F)Cl

Specifications

Color Yellow
Quantity 5 g
UN Number 1760
Formula Weight 215.00
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Dichlorobenzotrifluoride
RTECSNumber CZ5553755
TSCA No