accessibility menu, dialog, popup

UserName

CAS RN 320-50-3

IUPAC Name: 1,4-dichloro-2-(trifluoromethyl)benzene
Synonyms: 1,4-dichloro-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 215
Molecular Formula: C7H3Cl2F3
InChi Key: DYBYUWVMLBBEMA-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Cl)=CC=C1Cl
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results