missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorobenzonitrile, 98%

Catalog No. AAA1414106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1414106 5 g
AAA1414114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1414106 Supplier Thermo Scientific Chemicals Supplier No. A1414106

CAS: 21663-61-6 | C7H3Cl2N | 172.008 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21663-61-6
Molecular Formula C7H3Cl2N
Molecular Weight (g/mol) 172.008
MDL Number MFCD00019741
InChI Key LNGWRTKJZCBXGT-UHFFFAOYSA-N
Synonym benzonitrile, 2,5-dichloro, 2,5-dichloro-benzonitrile, 2,5-dichlorobenzonitrite, 2,5-dichlorobenzenecarbonitrile, pubchem3687, acmc-209fn5, ksc490q4h, 2,5-dichlorobenzonitrile, attercop-chm at133057
PubChem CID 89000
IUPAC Name 2,5-dichlorobenzonitrile
SMILES C1=CC(=C(C=C1Cl)C#N)Cl

Specifications

Melting Point 128°C to 131°C
Quantity 5 g
UN Number UN3439
Beilstein 2045293
Formula Weight 172.01
Percent Purity 98%
Chemical Name or Material 2,5-Dichlorobenzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 244-518-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only