missing translation for 'onlineSavingsMsg'
Learn More

2',5'-Dichloroacetoacetanilide 98.0+%, TCI America™

Catalog No. D10095G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D10095G 5 g
D100925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D10095G Supplier TCI America Supplier No. D10095G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2044-72-6
Molecular Formula C10H9Cl2NO2
Molecular Weight (g/mol) 246.087
MDL Number MFCD00018520
InChI Key HLMZZYYGOKOOTA-UHFFFAOYSA-N
Synonym 2',5'-dichloroacetoacetanilide, n-2,5-dichlorophenyl-3-oxobutanamide, acetoacetic acid-2,5-dichloroanilide, butanamide, n-2,5-dichlorophenyl-3-oxo, acetoacetanilide, 2',5'-dichloro, acmc-1cg2h, 2,5-dichloroacetoacetanilide, acetoacetanilide,5'-dichloro, n-acetoacetyl-2,5-dichloroaniline, butanamide,5-dichlorophenyl-3-oxo
PubChem CID 74890
IUPAC Name N-(2,5-dichlorophenyl)-3-oxobutanamide
SMILES CC(=O)CC(=O)NC1=C(C=CC(=C1)Cl)Cl

Specifications

Melting Point 95°C
Color White-Yellow
Quantity 5 g
Formula Weight 246.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2′,5′-Dichloroacetoacetanilide
EINECSNumber (3)-0268
TSCA Yes