missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloro-1,4-phenylenediamine 98.0+%, TCI America™

Catalog No. D187325G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D187325G 25 g
D1873250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D187325G Supplier TCI America Supplier No. D187325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 20103-09-7
Molecular Formula C6H6Cl2N2
Molecular Weight (g/mol) 177.028
MDL Number MFCD00007902
InChI Key QAYVHDDEMLNVMO-UHFFFAOYSA-N
Synonym 2,5-dichloro-1,4-phenylenediamine, 2,5-dichloro-p-phenylenediamine, 2,5-dichloro-1,4-benzenediamine, 1,4-diamino-2,5-dichlorobenzene, 1,4-benzenediamine, 2,5-dichloro, unii-9ccw77baxp, 1,4-diamino-3,6-dichlorobenzene, p-phenylenediamine, 2,5-dichloro, 9ccw77baxp, benzene, 2,5-dichloro, 1,4-diamino
PubChem CID 29949
IUPAC Name 2,5-dichlorobenzene-1,4-diamine
SMILES C1=C(C(=CC(=C1Cl)N)Cl)N

Specifications

Melting Point 166°C
Quantity 25 g
Formula Weight 177.03
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dichloro-1,4-phenylenediamine
EINECSNumber (3)-0295
RTECSNumber SS9150000
TSCA Yes