missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromothiazole, 97%

Catalog No. AAH2690914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2690914 25 g
AAH2690906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2690914 Supplier Thermo Scientific Chemicals Supplier No. H2690914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4175-78-4 | C3HBr2NS | 242.916 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4175-78-4
Molecular Formula C3HBr2NS
Molecular Weight (g/mol) 242.916
MDL Number MFCD00016891
InChI Key XIBIQFJKUZZLLX-UHFFFAOYSA-N
Synonym 2,5-dibromothiazole, thiazole, 2,5-dibromo, 2,5-dibormothiazole, thiazole,5-dibromo, 2,5-dibromo-thiazole, pubchem20949, acmc-209jlm, ksc237s3r, #
PubChem CID 312394
IUPAC Name 2,5-dibromo-1,3-thiazole
SMILES C1=C(SC(=N1)Br)Br

Specifications

Melting Point 47°C to 49°C
Quantity 25 g
Formula Weight 242.92
Percent Purity 97%
Chemical Name or Material 2,5-Dibromothiazole
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only