missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromoaniline, 97%

Catalog No. AAA1911422
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1911422 100 g
AAA1911406 5 g
AAA1911414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1911422 Supplier Thermo Scientific Chemicals Supplier No. A1911422
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3638-73-1 | C6H5Br2N | 250.921 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3638-73-1
Molecular Formula C6H5Br2N
Molecular Weight (g/mol) 250.921
MDL Number MFCD00007636
InChI Key WRTAZRGRFBCKBU-UHFFFAOYSA-N
Synonym 2,5-dibromobenzenamine, benzenamine, 2,5-dibromo, 2,5-dibromophenylamine, rarechem fh 1w 0043, pubchem3452, 2,5-dibromo-phenylamine, aniline, 2,5-dibromo, 2,5-bis bromanyl aniline, acmc-209im6, 2,5-dibromoaniline
PubChem CID 77198
IUPAC Name 2,5-dibromoaniline
SMILES C1=CC(=C(C=C1Br)N)Br

Specifications

Melting Point 51°C to 56°C
Flash Point >110°C (230°F)
Odor Characteristic
Quantity 100 g
UN Number UN2811
Beilstein 1817548
Formula Weight 250.93
Percent Purity 98%
Chemical Name or Material 2,5-Dibromoaniline
Hazard Category H301+H311+H331-H315-H319-H335-H373
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P314-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 222-865-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only