missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromo-3-methylthiophene 98.0+%, TCI America™

Catalog No. D43045G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43045G 5 g
D430425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43045G Supplier TCI America Supplier No. D43045G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13191-36-1
Molecular Formula C5H4Br2S
Molecular Weight (g/mol) 255.955
MDL Number MFCD00015470
InChI Key IHFXZROPBCBLLG-UHFFFAOYSA-N
Synonym thiophene, 2,5-dibromo-3-methyl, 2,5-dibromo-3-methyl-thiophene, acmc-1bt3y, 2-p-nitrophenylthio ethanol, 2,5-dibrom-3-methylthiophen, 3-methyl-2,5-dibromothiophene, 2,5-dibromo-3-methylthiophene
PubChem CID 83219
IUPAC Name 2,5-dibromo-3-methylthiophene
SMILES CC1=C(SC(=C1)Br)Br

Specifications

Color Yellow
Boiling Point 108°C
Quantity 5 g
Formula Weight 255.96
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Dibromo-3-methylthiophene
TSCA No
Recommended Storage Refrigerator