missing translation for 'onlineSavingsMsg'
Learn More

2,5-Diaminotoluene Dihydrochloride 98.0+%, TCI America™

Catalog No. D012625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D012625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D012625G Supplier TCI America Supplier No. D012625G
Only null left

Chemical Identifiers

CAS 615-45-2
Molecular Formula C7H12Cl2N2
Molecular Weight (g/mol) 195.09
MDL Number MFCD00035512
InChI Key VQUHVWVGRKTIBH-UHFFFAOYSA-N
Synonym 2,5-Toluenediamine Dihydrochloride, Tolylene-2,5-diamine Dihydrochloride, 2-Methyl-1,4-phenylenediamine Dihydrochloride
PubChem CID 11996
IUPAC Name dihydrogen 2-methylbenzene-1,4-diamine dichloride
SMILES [H+].[H+].[Cl-].[Cl-].CC1=CC(N)=CC=C1N

Specifications

Quantity 25 g
UN Number 2811
Formula Weight 195.09
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Diaminotoluene Dihydrochloride
EINECSNumber (3)-0126
RTECSNumber XT0350000
TSCA Yes