missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Diaminopyridine, 97%, ACROS Organics™
Supplier: Thermo Scientific Chemicals 360690250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| 2, 5-Diaminopyridine | |
| 180.0°C (12.0 mmHg) | |
| Glass bottle | |
| 25g | |
| MIROPXUFDXCYLG-UHFFFAOYSA-N | |
| pyridine-2,5-diamine | |
| 20314 | |
| 97% |
| 4318-76-7 | |
| 98% min. (HPLC) | |
| C5H7N3 | |
| 2,5-diaminopyridine, 2,5-pyridinediamine, pyridine, 2,5-diamino, diaminopyridine van, unii-h6olz060hi, h6olz060hi, da1, pyridine,5-diamino, 2,5-diamino pyridine, 2,5-diamino-pyridine | |
| C1=CC(=NC=C1N)N | |
| 109.13 | |
| 109.13 |
Safety and Handling
GHS Signal Word: Danger