missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Diamino-1,4-benzenedithiol Dihydrochloride 97.0+%, TCI America™
Supplier: TCI America D202225G
Specifications
| 2,5-Diamino-1,4-benzenedithiol Dihydrochloride | |
| 210°C | |
| C6H10Cl2N2S2 | |
| 25 g | |
| HVXLKRWRWNFGBA-UHFFFAOYSA-N | |
| 2,5-diaminobenzene-1,4-dithiol dihydrochloride | |
| 14618269 | |
| ≥97.0% (HPLC) |
| 75464-52-7 | |
| Yellow | |
| MFCD00142835 | |
| 2,5-Dimercapto-1,4-phenylenediamine Dihydrochloride | |
| Cl.Cl.NC1=CC(S)=C(N)C=C1S | |
| 245.18 | |
| 245.18 | |
| Crystalline Powder |
Chemical Identifiers
| 75464-52-7 | |
| 245.18 | |
| HVXLKRWRWNFGBA-UHFFFAOYSA-N | |
| 14618269 | |
| Cl.Cl.NC1=CC(S)=C(N)C=C1S |
| C6H10Cl2N2S2 | |
| MFCD00142835 | |
| 2,5-Dimercapto-1,4-phenylenediamine Dihydrochloride | |
| 2,5-diaminobenzene-1,4-dithiol dihydrochloride |
Safety and Handling
TSCA : No