missing translation for 'onlineSavingsMsg'
Learn More

2,5-Diamino-1,4-benzenedithiol Dihydrochloride 97.0+%, TCI America™

Catalog No. p-7127072
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D20225G 5 g
D202225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D20225G Supplier TCI America Supplier No. D20225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 75464-52-7
Molecular Formula C6H10Cl2N2S2
Molecular Weight (g/mol) 245.18
MDL Number MFCD00142835
InChI Key HVXLKRWRWNFGBA-UHFFFAOYSA-N
Synonym 2,5-Dimercapto-1,4-phenylenediamine Dihydrochloride
PubChem CID 14618269
IUPAC Name 2,5-diaminobenzene-1,4-dithiol dihydrochloride
SMILES Cl.Cl.NC1=CC(S)=C(N)C=C1S

Specifications

Melting Point 210°C
Color Yellow
Quantity 5 g
Formula Weight 245.18
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Diamino-1,4-benzenedithiol Dihydrochloride