missing translation for 'onlineSavingsMsg'
Learn More

2,5-Di(2,2,2-trifluoroethoxy)benzene-1-sulfonyl chloride, 97%, Thermo Scientific™

Numéro de catalogue. p-3881278
Modifier l'affichage
Click to view available options
Quantité:
1g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CD11334DA 1g
CD11334EA 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CD11334DA Fournisseur Thermo Scientific Chemicals Code fournisseur CD11334DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 152457-95-9
Formule moléculaire C10H7ClF6O4S
Poids moléculaire (g/mol) 372.66
Numéro MDL MFCD00052841
Clé InChI UCHAULDYIPVTOP-UHFFFAOYSA-N
Synonyme 2,5-bis 2,2,2-trifluoroethoxy benzenesulfonyl chloride, 2,5-bis 2,2,2-trifluoroethoxy benzene-1-sulfonyl chloride, 2,5-bis 2,2,2-trifluoroethoxy benzenesulfonylchloride, 2,5-bis 2,2,2-trifluoroethoxy benzenesulphonyl chloride, 2,5-bis 2,2,2-trifluoroethoxy benzenesulfonyl, 2,5-bis-2,2,2-trifluoroethoxy benzenesulfonyl chloride, benzenesulfonylchloride, 2,5-bis 2,2,2-trifluoroethoxy, 2,5-bis 2,2,2-trifluoroethoxy benzenesulphonyl chloride, tech., pubchem5052, acmc-1c3ia
PubChem CID 2736077
Nom de l’IUPAC 2,5-bis(2,2,2-trifluoroethoxy)benzenesulfonyl chloride
SOURIRES FC(F)(F)COC1=CC(=C(OCC(F)(F)F)C=C1)S(Cl)(=O)=O

Spécifications

Nom ou substance chimique 2,5-Di(2,2,2-trifluoroethoxy)benzene-1-sulfonyl chloride
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber Glass Bottle
Quantité 1g
Poids de formule 372.67
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.