missing translation for 'onlineSavingsMsg'
Learn More

2-(5-Bromo-2-thienyl)pyridine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC02210EB

Quantity 25g

 View more versions of this product

Catalog No. CC02210EB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

123784-07-6
240.12
AKNKWLQVFAYRRO-UHFFFAOYSA-N
736483
BrC1=CC=C(S1)C1=CC=CC=N1
C9H6BrNS
MFCD00120466
2-5-bromo-2-thienyl pyridine, 2-5-bromothiophen-2-yl pyridine, 2-5-bromothien-2-yl pyridine, pyridine,2-5-bromo-2-thienyl, acmc-20mqrz, 5-bromo-2-2-pyridyl thiophene, 2-5-bromo-2-thiophenyl pyridine, 2-bromo-5-2-pyridinyl thiophene, 2-5-bromanylthiophen-2-yl pyridine, 2-5-bromo-thiophen-2-yl-pyridine
2-(5-bromothiophen-2-yl)pyridine

Specifications

2-(5-Bromo-2-thienyl)pyridine
94.5
95%
25g
2-5-bromo-2-thienyl pyridine, 2-5-bromothiophen-2-yl pyridine, 2-5-bromothien-2-yl pyridine, pyridine,2-5-bromo-2-thienyl, acmc-20mqrz, 5-bromo-2-2-pyridyl thiophene, 2-5-bromo-2-thiophenyl pyridine, 2-bromo-5-2-pyridinyl thiophene, 2-5-bromanylthiophen-2-yl pyridine, 2-5-bromo-thiophen-2-yl-pyridine
BrC1=CC=C(S1)C1=CC=CC=N1
240.12
240.12
123784-07-6
100.0
C9H6BrNS
MFCD00120466
AKNKWLQVFAYRRO-UHFFFAOYSA-N
2-(5-bromothiophen-2-yl)pyridine
736483
95%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions