missing translation for 'onlineSavingsMsg'
Learn More

2-(4-tert-Butylphenyl)-5-(4-biphenylyl)-1,3,4-oxadiazole 98.0+%, TCI America™

Catalog No. B17675G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B17675G Supplier TCI America Supplier No. B17675G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15082-28-7
Molecular Formula C24H22N2O
Molecular Weight (g/mol) 354.453
MDL Number MFCD00003101
InChI Key XZCJVWCMJYNSQO-UHFFFAOYSA-N
Synonym butyl pbd, butyl-pbd, 2-4-biphenylyl-5-4-tert-butylphenyl-1,3,4-oxadiazole, 2-4-tert-butylphenyl-5-4-biphenylyl-1,3,4-oxadiazole, 2-1,1'-biphenyl-4-yl-5-4-tert-butyl phenyl-1,3,4-oxadiazole, 2-4-tert-butylphenyl-5-4-biphenyl-1,3,4-oxadiazole, 1,3,4-oxadiazole, 2-1,1'-biphenyl-4-yl-5-4-1,1-dimethylethyl phenyl, 2-biphenyl-4-yl-5-4-tert-butylphenyl-1,3,4-oxadiazole, 1,3,4-oxadiazole, 2-4-biphenylyl-5-p-tert-butylphenyl, 2-t-butylphenyl-5-4-biphenylyl-1,3,4-oxadiazole
PubChem CID 84782
IUPAC Name 2-(4-tert-butylphenyl)-5-(4-phenylphenyl)-1,3,4-oxadiazole
SMILES CC(C)(C)C1=CC=C(C=C1)C2=NN=C(O2)C3=CC=C(C=C3)C4=CC=CC=C4

Specifications

Melting Point 137°C
Color White-Yellow
Quantity 5 g
Formula Weight 354.45
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-tert-Butylphenyl)-5-(4-biphenylyl)-1,3,4-oxadiazole