missing translation for 'onlineSavingsMsg'
Learn More

acide 2-(4-pyridyl)-1,3-thiazole-4-carboxylique, Thermo Scientific™

Numéro de catalogue. CC34701DA
Modifier l'affichage
Click to view available options
Quantité:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC34701DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CC34701DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC34701DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 21278-86-4
Formule moléculaire C9H5N2O2S
Poids moléculaire (g/mol) 205.21
Numéro MDL MFCD00171745
Clé InChI COOQMBOJAAZEIR-UHFFFAOYSA-M
Synonyme 2-pyridin-4-yl thiazole-4-carboxylic acid, 2-4-pyridyl thiazole-4-carboxylic acid, 2-pyridin-4-yl-1,3-thiazole-4-carboxylic acid, 2-4-pyridyl-1,3-thiazole-4-carboxylic acid, 2-4-pyridinyl-1,3-thiazole-4-carboxylic acid, 2-pyridin-4-yl-thiazole-4-carboxylic acid, 4-thiazolecarboxylicacid, 2-4-pyridinyl, acmc-1cr1v, 4-thiazolecarboxylic acid, 2-4-pyridinyl, 4-carboxy-2-4-pyridyl thiazole
PubChem CID 716091
Nom de l’IUPAC acide 2-pyridine-4-yl-1,3-thiazole-4-carboxylique
SOURIRES [O-]C(=O)C1=CSC(=N1)C1=CC=NC=C1

Spécifications

Nom ou substance chimique 2-(4-pyridyl)-1, 3-thiazole-4-carboxylic Acid
CAS % minimum 94.5
CAS % maximal 100.0
Emballage Bouteille en verre ambrée
Quantité 1 g
Poids de formule 206.22
Pourcentage de pureté 95%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.