missing translation for 'onlineSavingsMsg'
Learn More

2,4-Pentanediol, (+/-) + meso, 99%

Catalog No. AAL0177909
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0177909 10 g
AAL0177918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0177909 Supplier Thermo Scientific Chemicals Supplier No. L0177909
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 625-69-4 | C5H12O2 | 104.149 g/mol

2,4-Pentanediol was used in the synthesis of chelated multinuclear complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Pentanediol was used in the synthesis of chelated multinuclear complexes.

Solubility
Fully miscible.

Notes
Hygroscopic. Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 625-69-4
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.149
MDL Number MFCD00004549
InChI Key GTCCGKPBSJZVRZ-UHFFFAOYSA-N
Synonym 2,4-pentanediol, 2,4-amylene glycol, isoamylene alcohol, pentanediol-2,4, 2,4-amyleneglycol, dl-2,4-pentanediol, 2,4-pentanediol dl+meso, wln: qy&1yq, 2,4-pentandiol, 2,4-dihydroxypentane
PubChem CID 12262
IUPAC Name pentane-2,4-diol
SMILES CC(CC(C)O)O

Specifications

Density 0.955
Boiling Point 200°C to 201°C
Flash Point 101°C (213°F)
Odor Odorless
Refractive Index 1.435
Quantity 10 g
Beilstein 969186
Sensitivity Hygroscopic
Solubility Information Fully miscible.
Formula Weight 104.15
Percent Purity 99%
Chemical Name or Material 2,4-Pentanediol, (+/-) + meso
Show More Show Less
EINECSNumber 210-907-5
RTECSNumber SA0490000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only